Assay Details
| Gene Symbol: spn-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072316.6 | NP_504717.1 | 4 - 5 | 950 | 101 | |
| GenBank mRNA | AB052819.1 | - | 4 - 5 | 953 | 101 | |
More Information
Set Membership: |
|

spn-4
Genomic Map