Assay Details
Gene Symbol: spe-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383328.1 | 2 - 3 | 297 | 96 | ||
NM_072317.7 | NP_504718.1 | 2 - 3 | 310 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
