Assay Details
Gene Symbol: gck-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072320.5 | NP_504721.1 | 6 - 7 | 1869 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
