Assay Details
| Gene Symbol: hpo-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072339.8 | NP_504740.2 | 4 - 5 | 350 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hpo-8| Gene Symbol: hpo-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072339.8 | NP_504740.2 | 4 - 5 | 350 | 68 | |
Set Membership: |
|