Assay Details
Gene Symbol: mys-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072395.4 | NP_504796.1 | 1 - 2 | 154 | 64 | |
GenBank mRNA | AY551963.1 | - | 1 - 2 | 175 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
