Assay Details
| Gene Symbol: uvs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072611.6 | NP_505012.2 | 2 - 3 | 707 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

uvs-1| Gene Symbol: uvs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072611.6 | NP_505012.2 | 2 - 3 | 707 | 76 | |
Set Membership: |
|