Assay Details
Gene Symbol: pals-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072646.3 | NP_505047.2 | 2 - 3 | 433 | 116 |
Gene Symbol: F26D11.12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129458.3 | NP_001122930.1 | 1 - 2 | 76 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001129458.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
