Assay Details
Gene Symbol: gpa-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072756.5 | NP_505157.1 | 5 - 6 | 477 | 119 | |
GenBank mRNA | AY008125.1 | - | 5 - 6 | 468 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
