Assay Details
Gene Symbol: lmd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373062.3 | NP_001360511.1 | 13 - 14 | 1998 | 69 | |
NM_001373063.2 | NP_001360537.1 | 12 - 13 | 1959 | 69 | ||
NM_001373064.1 | NP_001360538.1 | 12 - 13 | 1965 | 69 | ||
NM_072772.4 | NP_505173.2 | 13 - 14 | 2022 | 69 | ||
NM_072773.7 | NP_505174.3 | 13 - 14 | 2028 | 69 | ||
NM_072774.9 | NP_505175.2 | 14 - 15 | 2710 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
