Assay Details
Gene Symbol: lmd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373062.3 | NP_001360511.1 | 15 - 16 | 2383 | 113 | |
NM_001373063.2 | NP_001360537.1 | 14 - 15 | 2344 | 113 | ||
NM_001373064.1 | NP_001360538.1 | 14 - 15 | 2350 | 113 | ||
NM_072772.4 | NP_505173.2 | 15 - 16 | 2407 | 113 | ||
NM_072773.7 | NP_505174.3 | 15 - 16 | 2413 | 113 | ||
NM_072774.9 | NP_505175.2 | 16 - 17 | 3095 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
