Assay Details
Gene Symbol: lmd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072774.9 | NP_505175.2 | 13 - 14 | 2554 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
