Assay Details
Gene Symbol: gck-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028970.3 | NP_001024141.1 | 3 - 4 | 598 | 83 | |
NM_001313268.2 | NP_001300197.1 | 3 - 4 | 867 | 83 | ||
NM_001383362.1 | 3 - 4 | 482 | 83 | |||
NM_072909.5 | NP_505310.2 | 3 - 4 | 924 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
