Assay Details
Gene Symbol: gck-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028970.3 | NP_001024141.1 | 8 - 9 | 2029 | 137 | |
NM_001313268.2 | NP_001300197.1 | 8 - 9 | 2298 | 137 | ||
NM_001313269.3 | NP_001300198.1 | 8 | 1898 | 137 | ||
NM_001313270.4 | NP_001300199.1 | 8 - 9 | 2026 | 137 | ||
NM_001313271.4 | NP_001300200.1 | 9 | 1948 | 137 | ||
NM_001383362.1 | 8 - 9 | 1913 | 137 | |||
NM_072909.5 | NP_505310.2 | 8 - 9 | 2355 | 137 | ||
NM_072910.8 | NP_505311.3 | 8 - 9 | 2352 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
