Assay Details
Gene Symbol: aat-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072993.6 | NP_505394.2 | 2 - 3 | 729 | 195 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
