Assay Details
| Gene Symbol: fshr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073147.7 | NP_505548.1 | 11 - 12 | 2382 | 72 | |
| GenBank mRNA | AF224743.1 | - | 11 - 12 | 2261 | 72 | |
More Information
Set Membership: |
|

fshr-1
Genomic Map