Assay Details
| Gene Symbol: fshr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276924.3 | NP_001263853.1 | 3 - 4 | 599 | 131 | |
| NM_073147.7 | NP_505548.1 | 3 - 4 | 597 | 131 | ||
| GenBank mRNA | AF224743.1 | - | 3 - 4 | 476 | 131 | |
More Information
Set Membership: |
|

fshr-1
Genomic Map