Assay Details
Gene Symbol: fshr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276924.3 | NP_001263853.1 | 4 - 5 | 824 | 152 | |
NM_073147.7 | NP_505548.1 | 4 - 5 | 822 | 152 | ||
GenBank mRNA | AF224743.1 | - | 4 - 5 | 701 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
