Assay Details
| Gene Symbol: fshr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276924.3 | NP_001263853.1 | 9 - 10 | 1742 | 135 | |
| NM_073147.7 | NP_505548.1 | 9 - 10 | 1740 | 135 | ||
| GenBank mRNA | AF224743.1 | - | 9 - 10 | 1619 | 135 | |
More Information
Set Membership: |
|

fshr-1
Genomic Map