Assay Details
| Gene Symbol: rop-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073237.9 | NP_505638.1 | 1 - 2 | 80 | 71 | |
| GenBank mRNA | L41729.1 | - | 1 - 2 | 89 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rop-1| Gene Symbol: rop-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073237.9 | NP_505638.1 | 1 - 2 | 80 | 71 | |
| GenBank mRNA | L41729.1 | - | 1 - 2 | 89 | 71 | |
Set Membership: |
|