Assay Details
| Gene Symbol: rop-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073237.9 | NP_505638.1 | 2 - 3 | 987 | 78 | |
| GenBank mRNA | L41729.1 | - | 2 - 3 | 996 | 78 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  Detect Genomic DNA  Caenorhabditis elegans  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 rop-1
rop-1 Genomic Map
Genomic Map Back To Top
 Back To Top