Assay Details
| Gene Symbol: rop-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073237.9 | NP_505638.1 | 3 - 4 | 1646 | 145 | |
| GenBank mRNA | L41729.1 | - | 3 - 4 | 1655 | 145 | |
More Information
Set Membership: |
|

rop-1
Genomic Map