Assay Details
| Gene Symbol: ric-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129561.4 | NP_001123033.1 | 2 - 3 | 330 | 153 | |
| NM_073240.8 | NP_505641.2 | 2 - 3 | 199 | 153 | ||
More Information
Set Membership: |
|

ric-4
Genomic Map