Assay Details
Gene Symbol: che-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269275.3 | NP_001256204.1 | 14 - 15 | 2377 | 136 | |
NM_001269276.3 | NP_001256205.1 | 5 - 6 | 762 | 136 | ||
NM_001269277.3 | NP_001256206.1 | 3 - 4 | 305 | 136 | ||
NM_001307803.4 | NP_001294732.1 | 3 - 4 | 345 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
