Assay Details
Gene Symbol: che-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269275.3 | NP_001256204.1 | 16 - 17 | 2684 | 93 | |
NM_001269276.3 | NP_001256205.1 | 7 - 8 | 1069 | 93 | ||
NM_001269277.3 | NP_001256206.1 | 5 - 6 | 612 | 93 | ||
NM_001307803.4 | NP_001294732.1 | 5 - 6 | 652 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
