Assay Details
Gene Symbol: che-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269275.3 | NP_001256204.1 | 22 - 23 | 3785 | 128 | |
NM_001269276.3 | NP_001256205.1 | 13 - 14 | 2170 | 128 | ||
NM_001269277.3 | NP_001256206.1 | 11 - 12 | 1713 | 128 | ||
NM_001307803.4 | NP_001294732.1 | 11 - 12 | 1753 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
