Assay Details
Gene Symbol: che-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269275.3 | NP_001256204.1 | 8 - 9 | 1367 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
