Assay Details
| Gene Symbol: trx-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269278.2 | NP_001256207.1 | 1 - 2 | 144 | 86 | |
| NM_001269279.3 | NP_001256208.1 | 1 - 2 | 66 | 86 | ||
More Information
Set Membership: |
|

trx-2
Genomic Map