Assay Details
| Gene Symbol: sri-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001355085.1 | NP_001342025.1 | 2 - 3 | 288 | 99 | |
| NM_001373097.1 | NP_001359573.1 | 2 - 3 | 315 | 99 | ||
More Information
Set Membership: |
|

sri-20
Genomic Map