Assay Details
| Gene Symbol: sri-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001355085.1 | NP_001342025.1 | 4 - 5 | 831 | 96 | |
| NM_001373097.1 | NP_001359573.1 | 4 - 5 | 858 | 96 | ||
More Information
Set Membership: |
|

sri-20
Genomic Map