Assay Details
| Gene Symbol: ugt-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073265.4 | NP_505666.2 | 5 - 6 | 1009 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ugt-49| Gene Symbol: ugt-49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073265.4 | NP_505666.2 | 5 - 6 | 1009 | 113 | |
Set Membership: |
|