Assay Details
| Gene Symbol: ugt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073271.3 | NP_505672.2 | 7 - 8 | 1054 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ugt-2| Gene Symbol: ugt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073271.3 | NP_505672.2 | 7 - 8 | 1054 | 87 | |
Set Membership: |
|