Assay Details
Gene Symbol: nrx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129396.5 | NP_001122868.2 | 12 - 13 | 2180 | 89 | |
NM_001269333.4 | NP_001256262.1 | 12 - 13 | 2183 | 89 | ||
NM_001276930.3 | NP_001263859.1 | 13 - 14 | 2197 | 89 | ||
NM_001276931.3 | NP_001263860.1 | 13 - 14 | 2197 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
