Assay Details
Gene Symbol: nrx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129396.5 | NP_001122868.2 | 25 - 26 | 4782 | 65 | |
NM_001269333.4 | NP_001256262.1 | 24 - 25 | 4257 | 65 | ||
NM_001269334.3 | NP_001256263.1 | 5 - 6 | 854 | 65 | ||
NM_001269335.3 | NP_001256264.1 | 4 - 5 | 326 | 65 | ||
NM_001269336.2 | NP_001256265.1 | 4 - 5 | 1034 | 65 | ||
NM_001269337.3 | NP_001256266.1 | 4 - 5 | 626 | 65 | ||
NM_001276930.3 | NP_001263859.1 | 26 - 27 | 4799 | 65 | ||
NM_001276931.3 | NP_001263860.1 | 25 - 26 | 4271 | 65 | ||
NM_001307745.4 | NP_001294674.1 | 3 - 4 | 681 | 65 | ||
NM_001377602.2 | NP_001364673.1 | 2 - 3 | 296 | 65 | ||
GenBank mRNA | KF227795.1 | - | 3 - 4 | 485 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
