Assay Details
| Gene Symbol: eef-1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047635.6 | NP_001041100.1 | 2 - 3 | 185 | 91 | |
| NM_073399.6 | NP_505800.1 | 2 - 3 | 276 | 91 | ||
More Information
Set Membership: |
|

eef-1G
Genomic Map