Assay Details
| Gene Symbol: scb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383400.1 | 4 - 5 | 734 | 101 | ||
| Gene Symbol: mboa-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269372.3 | NP_001256301.2 | 4 - 5 | 441 | 101 | |
| GenBank mRNA | AF221132.1 | - | 4 - 5 | 422 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383400.1 transcripts.
Set Membership: |
|

scb-1
Genomic Map