Assay Details
| Gene Symbol: bgal-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073448.9 | NP_505849.2 | 2 - 3 | 315 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bgal-1| Gene Symbol: bgal-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073448.9 | NP_505849.2 | 2 - 3 | 315 | 102 | |
Set Membership: |
|