Assay Details
Gene Symbol: des-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269391.4 | NP_001256320.1 | 11 - 12 | 1889 | 58 | |
NM_001380763.2 | NP_001368114.1 | 8 - 9 | 1547 | 58 | ||
GenBank mRNA | AF077307.1 | - | 8 - 9 | 1562 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
