Assay Details
| Gene Symbol: mig-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073500.7 | NP_505901.2 | 7 - 8 | 1281 | 86 | |
| GenBank mRNA | AB044562.1 | - | 7 - 8 | 1262 | 86 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  Detect Genomic DNA  Caenorhabditis elegans  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 mig-17
mig-17 Genomic Map
Genomic Map Back To Top
 Back To Top