Assay Details
| Gene Symbol: ttll-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269402.3 | NP_001256331.1 | 4 - 5 | 870 | 62 | |
| NM_001269403.2 | NP_001256332.1 | 3 - 4 | 635 | 62 | ||
More Information
Set Membership: |
|

ttll-5
Genomic Map