Assay Details
Gene Symbol: bir-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073548.9 | NP_505949.1 | 2 - 3 | 188 | 65 | |
GenBank mRNA | U85911.1 | - | 2 - 3 | 146 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
