Assay Details
| Gene Symbol: mif-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380778.1 | 2 - 3 | 114 | 67 | ||
| NM_001380779.1 | NP_001366841.1 | 2 - 3 | 169 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mif-2| Gene Symbol: mif-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380778.1 | 2 - 3 | 114 | 67 | ||
| NM_001380779.1 | NP_001366841.1 | 2 - 3 | 169 | 67 | ||
Set Membership: |
|