Assay Details
| Gene Symbol: R07B7.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073628.6 | NP_506029.2 | 10 - 11 | 1413 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

R07B7.8| Gene Symbol: R07B7.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073628.6 | NP_506029.2 | 10 - 11 | 1413 | 88 | |
Set Membership: |
|