Assay Details
| Gene Symbol: R07B7.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073629.4 | NP_506030.2 | 4 - 5 | 1591 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

R07B7.10| Gene Symbol: R07B7.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073629.4 | NP_506030.2 | 4 - 5 | 1591 | 108 | |
Set Membership: |
|