Assay Details
| Gene Symbol: dpy-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073657.7 | NP_506058.1 | 1 - 2 | 51 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-30| Gene Symbol: dpy-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073657.7 | NP_506058.1 | 1 - 2 | 51 | 57 | |
Set Membership: |
|