Assay Details
| Gene Symbol: srpa-68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073682.7 | NP_506083.1 | 4 - 5 | 755 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srpa-68| Gene Symbol: srpa-68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073682.7 | NP_506083.1 | 4 - 5 | 755 | 75 | |
Set Membership: |
|