Assay Details
Gene Symbol: gcy-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073696.3 | NP_506097.3 | 3 - 4 | 344 | 161 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
