Assay Details
Gene Symbol: grl-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269502.3 | NP_001256431.1 | 6 - 7 | 669 | 77 | |
NM_001347271.3 | NP_001334227.1 | 2 - 3 | 310 | 77 | ||
NM_001347272.3 | NP_001334230.1 | 4 - 5 | 535 | 77 | ||
NM_001347273.3 | NP_001334228.1 | 5 - 6 | 568 | 77 | ||
NM_001347274.3 | NP_001334197.1 | 3 - 4 | 415 | 77 | ||
NM_001347275.3 | NP_001334229.1 | 5 - 6 | 634 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
