Assay Details
| Gene Symbol: cubn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073756.7 | NP_506157.3 | 6 - 7 | 1382 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cubn-1| Gene Symbol: cubn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073756.7 | NP_506157.3 | 6 - 7 | 1382 | 91 | |
Set Membership: |
|