Assay Details
| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313142.3 | NP_001300071.1 | 15 - 16 | 3306 | 96 | |
| NM_001313143.3 | NP_001300072.1 | 8 - 9 | 1341 | 96 | ||
| NM_073855.6 | NP_506256.3 | 19 - 20 | 4410 | 96 | ||
| GenBank mRNA | AY314773.1 | - | 19 - 20 | 4403 | 96 | |
More Information
Set Membership: |
|

fmi-1
Genomic Map