Assay Details
| Gene Symbol: fmi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313142.3 | NP_001300071.1 | 16 - 17 | 4628 | 93 | |
| NM_001313143.3 | NP_001300072.1 | 9 - 10 | 2663 | 93 | ||
| NM_073855.6 | NP_506256.3 | 20 - 21 | 5732 | 93 | ||
| GenBank mRNA | AY314773.1 | - | 20 - 21 | 5725 | 93 | |
More Information
Set Membership: |
|

fmi-1
Genomic Map